Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bossheads with hook

Stainless steel 18/10, anti-magnetic, 13 mm
  BOCH5304
 :   BOCHEM
Bossheads with hook
  BOCH5304
 :  BOCHEM
Bossheads with hook
Bossheads with hook

Stainless steel 18/10, anti-magnetic, 13 mm

 
23,30 €
   
1 * 1 items
 

Bossheads with hook, either made of stainless steel or nickel plated steel.

  • For rods up to 13 mm
  • Angle: 90°
 
  • Material:
    Stainless steel 18/10, anti-magnetic
  • For rod Ø:
    13 mm