Order Entry
Export
Orders LinkContactUsLinkComponent
Forceps for cover glasses

Forceps for cover glasses

Cover glass forceps, bent, PTFE coating, 105 mm
Catalog # 232-0021
Supplier:   BOCHEM
Forceps for cover glasses
Catalog # 232-0021
Supplier:  BOCHEM
Forceps for cover glasses
Forceps for cover glasses

Cover glass forceps, bent, PTFE coating, 105 mm

Price
29,20 €
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel or stainless steel with PTFE coating.

  • For easy, secure handling of cover glasses
  • With 6 mm wide tips
  • Can also be used for other laboratory equipment, such as filter discs
  • PTFE coated design with particularly smooth surface to prevent contamination and enable simple cleaning
Specifications
  • Description:
    Cover glass forceps, bent, PTFE coating
  • Length:
    105 mm