Orders LinkContactUsLinkComponent
Glass cutters
Glass cutters
Replacement hard metal blade
Catalog # BOCH12251
Supplier:   BOCHEM
Glass cutters
Catalog # BOCH12251
Supplier:  BOCHEM
Glass cutters
Glass cutters
Replacement hard metal blade
Price
51,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Plastic handle with hardened metal blade.
  • Clean and precise cutting
  • Replaceable hardened metal blades
  • Easy to change blade
Length 180 mm, blade dimensions 40×18 mm.
Specifications
  • Description:
    Replacement hard metal blade