Orders LinkContactUsLinkComponent
Forceps with guide pin, straight
Forceps with guide pin, straight
Sharp, straight, 145 mm
Catalog # BOCH1133
Supplier:   BOCHEM
Forceps with guide pin, straight
Catalog # BOCH1133
Supplier:  BOCHEM
Forceps with guide pin, straight
Forceps with guide pin, straight
Sharp, straight, 145 mm
Price
6,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Sharp, straight
  • Length:
    145 mm
  • Seq:
    04