3,4-Hexanedione 96%
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:4437-51-8
- Formula:C₆H₁₀O₂
- MDL Number:MFCD00010237
- Molecular Weight:114.14 g/mol
- Boiling Pt:129 °C (1013 hPa)
- Melting Pt:–10 °C
- Density:0.941 g/cm³
- Flash Pt:29 °C
- Storage Temperature:Ambient
- EINECS:224-651-7
- UN:1224
- ADR:3,III
Documents
ACRO29327
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4090 to 1.4150 (20°C, 589 nm)
- GC:≥95 %
- Specific gravity:(25°C) 0.93 to 0.95
Specifications
- Pack type:Glass bottle
- Pk:25 mL



