Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
19/26 NS, 19/26 NS, 140 mm
Catalog # 201-8160
Supplier:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Catalog # 201-8160
Supplier:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
19/26 NS, 19/26 NS, 140 mm
Price
78,30 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Total length:
    140 mm