Stirrer bearings with screw thread, SVL®
Stirrer bearing with Quickfit® ground glass con, SVL® stirrer shafts, 29/32, Borosilicate glass
Catalog # 201-8148
Supplier: SciLabware
Jump to:
Product Details & Documents
Specifications
- For:SVL® stirrer shafts
- Description:Stirrer bearing with Quickfit® ground glass con
- Material:Borosilicate glass
- Seq:04
- NS stopper:29/32