Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 40 mm
Catalog # 216-2770
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Excellent chemical resistance
- High temperature resistance
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifications
- Ø:40 mm
- Description:Watch glass, PYREX® borosilicate glass with firepolished edges
- Seq:01