Tetramethylsilane 99%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: TMS
- CAS Number:75-76-3
- Formula:C₄H₁₂Si
- MDL Number:MFCD00008274
- Molecular Weight:88.22 g/mol
- Boiling Pt:26.6 °C (1013 hPa)
- Melting Pt:–99 °C
- Density:0.64 g/cm³
- Flash Pt:–27 °C
- Storage Temperature:Refrigerator
- EINECS:200-899-1
- UN:2749
- ADR:3,I
Documents
ACRO26886
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3575 to 1.3595 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:500 mL



