n-Hexane ∼95% for HPLC
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
n-Hexane ∼95% for HPLC
Catalog # ACRO268360025
Supplier: Thermo Fisher Scientific
CAS Number: 92112-69-1
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:92112-69-1
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.659 g/cm³
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 %
- UV:at 200 nm A: ≤0.70
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0002 %
- Acidity:≤0.00015 meq/g
- Filtered to 0.2 micrometer:Filtered
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



