1,4-Dioxane 99.5% for HPLC
Glass bottle, 1 L
: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- :123-91-1
- :C₄H₈O₂
- :MFCD00006571
- :88.11 g/mol
- :13,03330
- :101.1 °C (1013 hPa)
- :12 °C
- :1.034 g/cm³
- :Ambient
- :204-661-8
- :1165
- :3,II
- Appearance:Clear colorless Liquid
- Water:≤0.025 %
- GC:≥99.4 %
- UV:at 218 nm A: ≤1
- Residue after evaporation:≤0.0002 %
- Filtered to 0.2 micrometer:Filtered
- Pack type:Glass bottle
- Pk:1 L



