Order Entry
Export
Orders LinkContactUsLinkComponent
Water, gradient grade for HPLC

Water, gradient grade for HPLC

Glass bottle, 1 L
Catalog # ACRO268300010
   Safer Human and Environmental Health
Water, gradient grade for HPLC
Catalog # ACRO268300010
   Safer Human and Environmental Health
CAS Number:  7732-18-5
Water, gradient grade for HPLC
Water, gradient grade for HPLC

Glass bottle, 1 L

CAS Number:  7732-18-5
Price
37,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Residue after evaporation:
    ≤0.0005 %
  • Residue after ignition:
    ≤0.0003 %
  • Gradient analysis:
    :
  • Absorbance largest eluted peak:
    ≤0.002 A.U.
  • Filtered to 0.2 micrometer:
    Filtered

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    ACRO268300010
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications