Orders LinkContactUsLinkComponent
Pipette Boxes, with Lid
Pipette Boxes, with Lid
Pipette box, for 64 pipettes of 5 ml / 36 shortie pipettes of 10 - 12 ml, 102×102×240 mm
Catalog # BELC1217-00409
Supplier:   BELLCO GLASS
Pipette Boxes, with Lid
Catalog # BELC1217-00409
Supplier:  BELLCO GLASS
Pipette Boxes, with Lid
Pipette Boxes, with Lid
Pipette box, for 64 pipettes of 5 ml / 36 shortie pipettes of 10 - 12 ml, 102×102×240 mm
Price
243,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
  • Stackable
  • Silicone pad on inside of top and bottom protect pipette tips
  • Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Specifications
  • Description:
    Pipette box, for 64 pipettes of 5 ml / 36 shortie pipettes of 10 - 12 ml
  • W×D×H:
    102×102×240 mm