2,3-Dichlorobenzaldehyde 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
2,3-Dichlorobenzaldehyde 99%
Catalog # ACRO232671000
Supplier: Thermo Fisher Scientific
CAS Number: 6334-18-5
Jump to:
Product Details & Documents
- CAS Number:6334-18-5
- Formula:C₇H₄Cl₂O
- MDL Number:MFCD00010127
- Molecular Weight:175.01 g/mol
- Boiling Pt:144 °C (30 mmHg)
- Melting Pt:60…64 °C
- Density:1,33 g/cm³ (100 °C)
- Flash Pt:135 °C
- Storage Temperature:Ambient
- EINECS:228-711-3
- UN:3261
- ADR:8,II
Documents
ACRO23267
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:60 to 64 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



