2,3-Dichlorobenzaldehyde 99%
Glass bottle for solids, 25 g
- :6334-18-5
- :C₇H₄Cl₂O
- :MFCD00010127
- :175.01 g/mol
- :144 °C (30 mmHg)
- :60…64 °C
- :1,33 g/cm³ (100 °C)
- :135 °C
- :Ambient
- :228-711-3
- :3261
- :8,II
- Appearance:White to light yellow Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:60 to 64 °C
- GC:≥98.5 %
- Pack type:Glass bottle for solids
- Pk:25 g



