Chloroform 99+% stabilised for analysis
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised for analysis
Catalog # ACRO232090010
Supplier: Thermo Fisher Scientific
CAS Number: 67-66-3
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61 °C (760 mmHg)
- Melting Pt:–63 °C
- Density:1.48 g/cm³
- EINECS:200-663-8
- ADR:6.1,III
- :Stabilised with ethanol
- Storage Temperature:Ambient
- UN:1888
- REACH:05-2114614906-45
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.01 %
- GC:≥99.0 % (Excluding stabilizer)
- Color scale:≤10 APHA
- Stabilizer:0.6 to 1.0 % ethanol
- Residue after evaporation:≤0.001 %
- Free chlorine:Passes test
- Cadmium (Cd):≤0.1 ppm
- Cobalt (Co):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Nickel (Ni):≤0.1 ppm
- Zinc (Zn):≤0.5 ppm
- Acidity (HCl):≤0.001 %
- Acetone and aldehyde:≤0.005 % (as Acetone)
Specifications
- Pack type:Glass bottle
- Pk:1 L



