(+)-Cinchonine 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
- CAS Number:118-10-5
- Formula:C₁₉H₂₂N₂O
- MDL Number:MFCD00064372
- Molecular Weight:294.4 g/mol
- Merck Index:13,02308
- Melting Pt:255…265 °C
- Density:1,2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-234-6
Documents
ACRO22781
SDS
Specifications
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:255 to 265 °C
- Loss on drying:≤1.0 %
- Sulfated ash:≤0.1 %
- Specific optical rotation:+251° to +269° (20 °C, 589 nm) (c=2, 0.2M HCl)
- Titration with HClO4:98.5 to 101.5 %
- Other alkaloids:≤3 % (HPLC) Quinidine + hydroquinidine
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



