Colchicine 97%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine
- CAS Number:64-86-8
- Formula:C₂₂H₂₅NO₆
- MDL Number:MFCD00078484
- Molecular Weight:399.44 g/mol
- Merck Index:12,02536
- Melting Pt:150…160 °C (dec.)
- Density:1,32 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-598-5
- UN:1544
- ADR:6.1,I
Documents
ACRO22712
SDS
Specifications
Specification Test Results
- Appearance:Light yellow Powder
- Infrared spectrum:Conforms
- Water:≤2 % (K.F.)
- HPLC:≥96.0 % on anhydrous and solvent-free substance
- Specific optical rotation:-240° to -250° (20°C, 589 nm) (c=1, alcohol) (on anhydrous and solvent-free substance
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



