Progesterone 98%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:57-83-0
- Formula:C₂₁H₃₀O₂
- MDL Number:MFCD00003658
- Melting Pt:126…131 °C
- Storage Temperature:Ambient
- EINECS:200-350-6
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:126 to 131 °C
- Loss on drying:≤0.5 % (500 mg, 105 °C, 2 hrs)
- HPLC:≥97.5 %
- Specific optical rotation:+186° to +194° (20 °C, 589 nm) (c=1, ethanol) (on dried substance)
Specifications
- Pack type:Glass bottle
- Pk:25 g



