Order Entry
Export
Orders LinkContactUsLinkComponent
Sample loops for Valco valves

Sample loops for Valco valves

Sample loop, PEEK, 1000 µl, max. 240 bar
Catalog # JOURJR-SLV-75-1000
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Catalog # JOURJR-SLV-75-1000
Supplier:  VICI JOUR RESEARCH
Sample loops for Valco valves

Sample loop, PEEK, 1000 µl, max. 240 bar

Price
147,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.

  • Fits Valco W and UW type valves
  • Tolerance of ±0,025 mm

PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.

Delivery information: Supplied with PEEK hex-head nuts and PEEK ferrules.

Specifications
  • Capacity:
    1000 µl
  • Description:
    Sample loop
  • Material:
    PEEK
  • Seq:
    08
  • Pressure:
    max. 240 bar