Orders LinkContactUsLinkComponent
Magnetic stirring bars, PTFE
Magnetic stirring bars, PTFE
Set with IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 and TRIKA® 25 and 40 stirring bars
Catalog # 441-2121
Supplier:   IKA
Magnetic stirring bars, PTFE
Catalog # 441-2121
Supplier:  IKA
Magnetic stirring bars, PTFE
Magnetic stirring bars, PTFE
Set with IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 and TRIKA® 25 and 40 stirring bars
Price
146,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Specifications
  • Type:
    Set with IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 and TRIKA® 25 and 40 stirring bars