Pipette tips for labopette®
1 - 250 µl, Transparent, labopette® multichannel pipettes, 10 racks, 96 each
Catalog # HIRS9490206
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifications
- Packed:10 racks, 96 each
- Volume:1 - 250 µl
- Seq:11
- Tip colour:Transparent
- For use with:labopette® multichannel pipettes