Pipette tips for labopette®
50 - 1200 µl, Transparent, Hirschmann Laborgeräte pipettes, 10 racks, 96 each
Catalog # HIRS9490205
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifications
- Packed:10 racks, 96 each
- Volume:50 - 1200 µl
- Seq:13
- Tip colour:Transparent
- For use with:Hirschmann Laborgeräte pipettes