Pipette tips for labopette®
0,2 - 10 µl, Transparent, Hirschmann Laborgeräte, Gilson® pipettes, 10 racks, 96 each
Catalog # HIRS9490201
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifications
- Packed:10 racks, 96 each
- Volume:0,2 - 10 µl
- Seq:09
- Tip colour:Transparent
- For use with:Hirschmann Laborgeräte, Gilson® pipettes