Pipette tips for labopette®
1 - 200 µl, Yellow, Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettes, Bulk packed, bag of 1000
Catalog # HIRS9490102
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifications
- Packed:Bulk packed, bag of 1000
- Volume:1 - 200 µl
- Seq:02
- Tip colour:Yellow
- For use with:Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson® pipettes