Order Entry
Export
Orders LinkContactUsLinkComponent
Digital burettes, solarus®

Digital burettes, solarus®

20 ml, 0,01 ml, ±0,2 %, <0,1 %
Catalog # 613-3255
Supplier:   HIRSCHMANN
Digital burettes, solarus®
Catalog # 613-3255
Supplier:  HIRSCHMANN
Digital burettes, solarus®
Digital burettes, solarus®

20 ml, 0,01 ml, ±0,2 %, <0,1 %

Price
1 050,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Variable volume.

  • Integral solar panel provides energy for electronic components such as the digital display, so batteries no longer need to be replaced or charged
  • Easy to use recirculation system for rapid priming with no loss of reagents
  • Bi-directional RS232 interface and USB
  • Parts that come into contact with liquids are autoclavable
  • Certificate of quality and conformity

These innovative digital burettes, with integrated photovoltaic cells and manual control, are designed for exact titration.

Ordering information: Supplied with three adapters (A32, A38, S40), a discharge tube, a filling tube with a locking screw, individual serial number and a test certificate.

Specifications
  • Capacity:
    20 ml
  • Accuracy:
    ±0,2 %
  • Division:
    0,01 ml
  • Imprecision:
    <0,1 %
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Individual certificate of performance
    Individual certificate of performance
    613-2938
    Log in for availability
    1 * 1 items - 28,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Charger, 100 - 240 V, EU plug
    Charger, 100 - 240 V, EU plug
    661-4219
    Log in for availability
    1 * 1 items - 49,30 €

Product Information

  • Catalog #
    Availability
    Price
  • Amber glass bottle, 2500 ml, round
    Amber glass bottle, 2500 ml, round
    661-4223
    Log in for availability
    1 * 1 items - 21,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Amber glass bottle, 500 ml
    Amber glass bottle, 500 ml
    612-2295
    Log in for availability
    1 * 1 items - 9,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube unit, Luer-Lock
    Discharge tube unit, Luer-Lock
    612-2290
    Log in for availability
    1 * 1 items - 69,10 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube unit, Luer-Lock
    Discharge tube unit, Luer-Lock
    612-2291
    Log in for availability
    1 * 1 items - 96,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube unit, complete
    Discharge tube unit, complete
    612-2288
    Log in for availability
    1 * 1 items - 26,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Fine ejection unit
    Fine ejection unit
    613-1658
    Log in for availability
    1 * 1 items - 34,30 €

Product Information

  • Catalog #
    Availability
    Price
  • Ground adapter, silicone, A 45 compliant with NS 29/32
    Ground adapter, silicone, A 45 compliant with NS 29/32
    613-1656
    Log in for availability
    1 * 1 items - 27,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Double stand sleeve, stainless steel
    Double stand sleeve, stainless steel
    613-3862
    Log in for availability
    1 * 1 items - 69,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter A45 with filling tube NS 12/21
    Adapter A45 with filling tube NS 12/21
    613-1655
    Log in for availability
    1 * 1 items - 225,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Base for separate media supply to enable suctioning of the medium directly from a separate storage container using a tube
    Base for separate media supply to enable suctioning of the medium directly from a separate storage container using a tube
    613-3861
    Log in for availability
    1 * 1 items - 293,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube unit, FEP
    Discharge tube unit, FEP
    612-0916
    Log in for availability
    1 * 1 items - 27,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Connecting cable RS232
    Connecting cable RS232
    612-2274
    Log in for availability
    1 * 1 items - 107,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, A 45 - S 40, PP
    Adapter, A 45 - S 40, PP
    612-0920
    Log in for availability
    1 * 1 items - 11,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, A 45 - A 38, PP
    Adapter, A 45 - A 38, PP
    612-0919
    Log in for availability
    1 * 1 items - 11,80 €

Product Information

  • Catalog #
    Availability
    Price
  • USP individual certificate of performance
    USP individual certificate of performance
    613-2939
    Log in for availability
    1 * 1 items - 28,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, A 45 - A 32, PP
    Adapter, A 45 - A 32, PP
    612-0918
    Log in for availability
    1 * 1 items - 11,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (100 filters)
    Round filter set with one-sided connection (100 filters)
    HIRS9903706
    Log in for availability
    1 * 100 items - 601,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (50 filters)
    Round filter set with one-sided connection (50 filters)
    HIRS9903705
    Log in for availability
    1 * 50 items - 347,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DKD calibration certificate for solarus® 20, 50 ml
    DKD calibration certificate for solarus® 20, 50 ml
    613-2265
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Ejection unit /discharge tube for variable titration
    Ejection unit /discharge tube for variable titration
    613-0894
    Log in for availability
    1 * 1 items - 51,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Ejection unit/discharge tube, flexible, spiral shaped, inner Ø 1,5 mm
    Ejection unit/discharge tube, flexible, spiral shaped, inner Ø 1,5 mm
    613-1654
    Log in for availability
    1 * 1 items - 72,40 €