Order Entry
Export
Orders LinkContactUsLinkComponent
Micro capillaries, disposable, minicaps

Micro capillaries, disposable, minicaps

Sodium-heparinised minicaps, 4 µl, 30 - 32 mm
Catalog # 720-2837
Supplier:   HIRSCHMANN
Micro capillaries, disposable, minicaps
Catalog # 720-2837
Supplier:  HIRSCHMANN
Micro capillaries, disposable, minicaps
Micro capillaries, disposable, minicaps

Sodium-heparinised minicaps, 4 µl, 30 - 32 mm

Price
111,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents

Disposable capillary pipettes with the end-to-end system. With genuine Schott DURAN® glass. Accuracy ±≤0,5%, CV ±≤1%. With batch number on the package.

  • Correct filling of the capillary
  • Conformity certified, according to the German 'Eichordnung' certification
  • Colour-coded vial cap with capillary dispenser
  • Safe packaging for protection during transportation

Packaging: 100 minicaps packed in a glass vial with dispenser cap. 10 vials plus a pipetting device are packed into a outer carton.

Specifications
  • Description:
    Sodium-heparinised minicaps, 4 µl
  • Length:
    30 - 32 mm
  • Seq:
    04