Stopcocks for burettes, lateral
50 ml burettes
HIRS8901175
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
- For:50 ml burettes
- Seq:03
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.