Orders LinkContactUsLinkComponent
Stopcocks for burettes, straight
Stopcocks for burettes, straight
50 ml burettes
Catalog # HIRS8900875
Supplier:   HIRSCHMANN
Stopcocks for burettes, straight
Catalog # HIRS8900875
Supplier:  HIRSCHMANN
Stopcocks for burettes, straight
Stopcocks for burettes, straight
50 ml burettes
Price
52,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Straight burette stopcock, made of borosilicate glass, with glass key for burettes, clear glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Specifications
  • For:
    50 ml burettes
  • Seq:
    03