Stopcocks for burettes, straight
2 - 10 ml burettes
HIRS8900860
Straight burette stopcock, made of borosilicate glass, with glass key for burettes, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
- For:2 - 10 ml burettes
- Seq:01