Counting chambers, Neubauer
Counting chamber with clamps, bright-lined, Neubauer
HIRS8110203
Counting chambers with double net ruling.
: Include two cover glasses.
- Description:Counting chamber with clamps, bright-lined, Neubauer
Counting chambers with double net ruling.
: Include two cover glasses.