Counting chambers, Neubauer Imp.
Counting chamber with double ruling, with clamps, Imp. Neubauer, 0,1 mm
Catalog # HIRS8110104
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Cell counting chambers designed with double net ruling.
Ordering information: Includes two cover glasses.
Documents
Specifications
- Depth of chamber:0,1 mm
- Description:Counting chamber with double ruling, with clamps, Imp. Neubauer