Orders LinkContactUsLinkComponent
Counting chambers, Neubauer
Counting chambers, Neubauer
Counting chamber with clamps, Neubauer
Catalog # HIRS8110103
Supplier:   HIRSCHMANN
Counting chambers, Neubauer
Catalog # HIRS8110103
Supplier:  HIRSCHMANN
Counting chambers, Neubauer
Counting chambers, Neubauer
Counting chamber with clamps, Neubauer
Price
96,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Counting chambers with double net ruling.
Ordering information: Include two cover glasses.
Specifications
  • Description:
    Counting chamber with clamps, Neubauer