Counting chambers, Bürker
Counting chamber with clips, Bürker, 0,1 mm
Catalog # 612-5727
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These counting chambers with double net ruling are suitable for in vitro diagnostic applications.
Certifications: In vitro Diagnostic Devices Directive (98/79/EC)
Ordering information: Includes two cover glasses.
Documents
Specifications
- Depth of chamber:0,1 mm
- Type:Bürker
- Description:Counting chamber with clips