Orders LinkContactUsLinkComponent
Counting chambers, Neubauer Imp.
Counting chambers, Neubauer Imp.
Counting chamber with double ruling, bright-lined, Imp. Neubauer, 0,1 mm
Catalog # HIRS8100204
Supplier:   HIRSCHMANN
Counting chambers, Neubauer Imp.
Catalog # HIRS8100204
Supplier:  HIRSCHMANN
Counting chambers, Neubauer Imp.
Counting chambers, Neubauer Imp.
Counting chamber with double ruling, bright-lined, Imp. Neubauer, 0,1 mm
Price
153,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Cell counting chambers designed with double net ruling.
Ordering information: Includes two cover glasses.
Specifications
  • Depth of chamber:
    0,1 mm
  • Description:
    Counting chamber with double ruling, bright-lined, Imp. Neubauer