Counting chambers, Bürker
Counting chamber, bright-lined, Bürker, 0,10 mm
Catalog # 612-5723
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These counting chambers with double net ruling are suitable for in vitro diagnostic applications.
Certifications: In vitro Diagnostic Devices Directive (98/79/EC)
Ordering information: Includes two cover glasses.
Documents
Specifications
- Depth of chamber:0,10 mm
- Type:Bürker
- Description:Counting chamber, bright-lined