Counting chambers, Thoma
Counting chamber, double ruling, Thoma, 0,10 mm
Catalog # 612-5721
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Cell counting chambers including two cover glasses for haemacytometers.
Specifications
- Depth of chamber:0,10 mm
- Description:Counting chamber, double ruling, Thoma