Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

180 mm, 14 mm, 120 mm
Catalog # HIRS7750107
Supplier:   HIRSCHMANN
Funnels with short stem
Catalog # HIRS7750107
Supplier:  HIRSCHMANN
Funnels with short stem
Funnels with short stem

180 mm, 14 mm, 120 mm

Price
46,10 €
Log in for availability
1 * 12 items
Product Details & Documents

Funnels, made of PP, translucent. With smooth interior surfaces.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifications
  • Funnel O-Ø:
    120 mm
  • Stem O-Ø:
    14 mm
  • Stem length:
    180 mm