Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

65 mm, 5 mm, 40 mm
Catalog # HIRS7750103
Supplier:   HIRSCHMANN
Funnels with short stem
Catalog # HIRS7750103
Supplier:  HIRSCHMANN
Funnels with short stem
Funnels with short stem

65 mm, 5 mm, 40 mm

Price
38,80 €
Log in for availability
1 * 24 items
Product Details & Documents

Funnels, made of PP, translucent. With smooth interior surfaces.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifications
  • Funnel O-Ø:
    40 mm
  • Stem O-Ø:
    5 mm
  • Stem length:
    65 mm