Order Entry
Export
Orders LinkContactUsLinkComponent
Bulb pipettes, two marks, class AS

Bulb pipettes, two marks, class AS

100 ml, 0,08 ± ml, 600 mm, -
Catalog # 612-2792
Supplier:   HIRSCHMANN
Bulb pipettes, two marks, class AS
Catalog # 612-2792
Supplier:  HIRSCHMANN
Bulb pipettes, two marks, class AS
Bulb pipettes, two marks, class AS

100 ml, 0,08 ± ml, 600 mm, -

Price
25,10 €
Log in for availability
1 * 1 items
Product Details & Documents

AR-Glas®

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in amber stain
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)

DIN 12691

 : DIN EN ISO 648; DE-M marking

Specifications
  • Capacity:
    100 ml
  • Length:
    600 mm
  • Suppl. to DIN:
    -
  • Tolerance:
    0,08 ± ml