Order Entry
Export
Orders LinkContactUsLinkComponent
Bulb pipettes, one mark, class B

Bulb pipettes, one mark, class B

10 ml, 0,03 ± ml, 440 mm, -
Catalog # 612-1352
Supplier:   HIRSCHMANN
Bulb pipettes, one mark, class B
Catalog # 612-1352
Supplier:  HIRSCHMANN
Bulb pipettes, one mark, class B
Bulb pipettes, one mark, class B

10 ml, 0,03 ± ml, 440 mm, -

Price
8,30 €
Log in for availability
1 * 1 items
Product Details & Documents

AR-Glas®

  • Calibrated to deliver (TD, Ex)
  • With silver diffusion ink durable brown graduations

DIN 12690

Certifications: DIN EN ISO 648; DE-M marking

Specifications
  • Capacity:
    10 ml
  • Length:
    440 mm
  • Suppl. to DIN:
    -
  • Tolerance:
    0,03 ± ml