Jars with base, knobbed lid
180 mm, 180 mm
Catalog # 216-4296
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made of glass, with glass or stainless steel lid.
- Good chemical resistance
- Sturdy
- Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Specifications
- Ext.Ø:180 mm
- Height:180 mm