Orders LinkContactUsLinkComponent
Jars with base, knobbed lid
Jars with base, knobbed lid
100 mm, 150 mm
Catalog # 216-4294
Jars with base, knobbed lid
Catalog # 216-4294
Jars with base, knobbed lid
Jars with base, knobbed lid
100 mm, 150 mm
Price
23,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Made of glass, with glass or stainless steel lid.
  • Good chemical resistance
  • Sturdy
  • Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Specifications
  • Ext.Ø:
    100 mm
  • Height:
    150 mm