Order Entry
Export
Orders LinkContactUsLinkComponent
Instrument trays

Instrument trays

200×100×100 mm
Catalog # HECH42910010
Instrument trays
Catalog # HECH42910010
Instrument trays
Instrument trays

200×100×100 mm

Price
67,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Glass with chrome-plated knobbed lid.

  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Good chemical resistance

Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.

Specifications
  • W×D×H:
    200×100×100 mm