Order Entry
Export
Orders LinkContactUsLinkComponent
Test Tubes, round bottom, with spout

Test Tubes, round bottom, with spout

5 ml, 110 mm, 12 mm
Catalog # 212-1602
Test Tubes, round bottom, with spout
Catalog # 212-1602
Test Tubes, round bottom, with spout
Test Tubes, round bottom, with spout

5 ml, 110 mm, 12 mm

Price
2,70 €
Log in for availability
1 * 1 items
Product Details & Documents

AR® (soda glass).

  • Fused white graduation
  • Good chemical and physical properties
  • Round bottom

Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.

Specifications
  • Ext.Ø:
    12 mm
  • Capacity:
    5 ml
  • Length:
    110 mm