Order Entry
Export
Orders LinkContactUsLinkComponent
Raschig rings

Raschig rings

Raschig ring, 10 mm, 10 mm
  HECH41400010
Raschig rings
  HECH41400010
Raschig rings
Raschig rings

Raschig ring, 10 mm, 10 mm

 
32,70 €
   
1 * 1 kg
 

Soda lime glass.

  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
 
  • Ext.Ø:
    10 mm
  • Description:
    Raschig ring
  • Height:
    10 mm
  • Seq:
    06