Cover glasses
#1, 25 mm
Catalog # 631-9509
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
: Available in different diameters.
Specifications
- Ø:25 mm
- No.:#1
- Seq:13