Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
#1, 12 mm
Catalog # HECH41001112
Cover glasses
Catalog # HECH41001112
Cover glasses
Cover glasses
#1, 12 mm
Price
91,50 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
 : Available in different diameters.
Specifications
  • Ø:
    12 mm
  • No.:
    #1
  • Seq:
    05