Order Entry
Export
Orders LinkContactUsLinkComponent
Crown stopwatches

Crown stopwatches

Analogue stopwatch, 1/10 s, 15 min, 120 g, Chrome, 55 mm
Catalog # 609-1128
Supplier:   HANHART
Crown stopwatches
Catalog # 609-1128
Supplier:  HANHART
Crown stopwatches
Crown stopwatches

Analogue stopwatch, 1/10 s, 15 min, 120 g, Chrome, 55 mm

Price
365,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Chrome plated and polished brass casing, diamond cut.

  • Pin lever, seven jewels
  • Protected against shock, dust and water
  • Industry model: Additional case seals to give extra protection against dust and water
Specifications
  • Ø:
    55 mm
  • Colour:
    Chrome
  • Description:
    Analogue stopwatch, 1/10 s, 15 min, 120 g