Knife spatulas, angled
205 mm, 105 mm
HAMMHSN236-10
Jump to:
Flexible knife spatulas with wooden handle and blade made of stainless steel.
Specifications
- Length:205 mm
- Seq:15
- Blade length:105 mm
Flexible knife spatulas with wooden handle and blade made of stainless steel.